Carboxymethylcellulose Natriumsalz

Lieferant: Thermo Fisher Scientific

Synonyme: Carboxymethyl cellulose, sodium salt, Cellulose gum, Cellulose, carboxymethyl ether, sodium salt, Sodium carboxymethyl cellulose (Viscosity 800mpa.s), Sodium carboxymethyl cellulose, CMC

A18105.0E A18105.36
A18105.0EEA 226 EUR
A18105.0E A18105.36
Carboxymethylcellulose Natriumsalz
Carboxymethyl cellulose sodium salt

CAS No.: 9004-32-4 Melting Point: 300° Merck Reference: 14,1829 MDL No.: MFCD00081472

Formel: [C₆H₇O₂(OH)x(OCH₂COONa)y]n
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00081472
CAS-Nummer: 9004-32-4
Merck Index: 14,1829

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR