1,4-Dibromtetrafluorbenzol ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

A18135.03 A18135.06 A18135.14
A18135.03EA 37.5 EUR
A18135.03 A18135.06 A18135.14
1,4-Dibromtetrafluorbenzol ≥99%
1,4-Dibromtetrafluorbenzol

Purity: 99% CAS No.: 344-03-6 Molecular Formula: C6Br2F4 Formula Weight: 307.87 Melting Point: 78-81° MDL No.: MFCD00000309

Formel: C₆Br₂F₄
Molecular Weight: 307.87 g/mol
Siedepunkt: 198
Schmelzpunkt: 78…81 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000309
CAS-Nummer: 344-03-6
EINECS: 206-448-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR