Cacodylsäure Natriumsalz Trihydrat ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Dimethylarsinic acid sodium salt trihydrate, Sodium cacodylate trihydrate, Dimethylarsonic acid sodium salt trihydrate, Sodium dimethylarsinate trihydrate

A18139.22 A18139.14 A18139.36
A18139.22EA 211 EUR
A18139.22 A18139.14 A18139.36
Cacodylsäure Natriumsalz Trihydrat ≥98%
Cacodylsäure Natriumsalz Trihydrat

Purity: 98+% CAS No.: 6131-99-3 Molecular Formula: C2H6AsNaO2.3H2O Formula Weight: 214.03 (159.98anhy) Beilstein Reference: 3702348 Merck Reference: 14,8595 MDL No.: MFCD00149079


An integral component of terminal deoxynucleotidyl transferase buffer; also useful in electron microscopy as a bactericide buffer

Formel: (CH₃)₂As(O)ONa·3H₂O
Molecular Weight: 213.94 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00149079
CAS-Nummer: 6131-99-3
EINECS: 204-708-2
UN: 1688
ADR: 6.1 II

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR