4-Chlor-2-nitrophenol, (max. 10% H₂O) ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A18150.14
A18150.14EA 31.2 EUR
A18150.14
4-Chlor-2-nitrophenol, (max. 10% H₂O) ≥98%
4-Chlor-2-nitrophenol

Purity: 98% CAS No.: 89-64-5 Molecular Formula: C6H4ClNO3 Formula Weight: 173.56 Melting Point: 85-88° Beilstein Reference: 2048031

Formel: C₆H₄ClNO₃
Schmelzpunkt: 85…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007113
CAS-Nummer: 89-64-5
EINECS: 201-927-5
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR