1,4-Dibromnaphthalin ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A18171.06 A18171.14
A18171.06EA 114 EUR
A18171.06 A18171.14
1,4-Dibromnaphthalin ≥98%
1,4-Dibromonaphthalene

Purity: 98% CAS No.: 83-53-4 Molecular Formula: C10H6Br2 Formula Weight: 285.97 Melting Point: 80-82° MDL No.: MFCD00041823

Formel: C₁₀H₆Br₂
Molecular Weight: 285.97 g/mol
Siedepunkt: 310
Schmelzpunkt: 80…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041823
CAS-Nummer: 83-53-4
EINECS: 201-484-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR