Nɑ-Benzoyl-L-arginine ethyl ester hydrochloride ≥98%

Lieferant: Thermo Scientific

Synonyme: Bz-Arg-OEt.HCl, Nα-Benzoyl-L-arginine Ethyl Ester HCl

A18181.06
A18181.06EA 65.2 EUR
A18181.06
Nɑ-Benzoyl-L-arginine ethyl ester hydrochloride ≥98%
Nalpha-Benzoyl-L-arginine ethyl ester hydrochloride

Purity: 98+% CAS No.: 2645-08-1 Molecular Formula: C15H22N4O3.HCl Formula Weight: 342.84 Storage Temperature: -30°C to -10°C Melting Point: 127-131° Beilstein Reference: 3781694 Optical Rotation: -17° (c=2, water) MDL No.: MFCD00012579

Formel: C₁₅H₂₂N₄O₃
Molecular Weight: 342,82 g/mol
Schmelzpunkt: 127…131 °C
MDL: MFCD00012579
CAS-Nummer: 2645-08-1
EINECS: 220-157-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR