3-Aminophenylboronsäure Monohydrat 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Dihydroxy(3-aminophenyl)boran Monohydrat, (3-Aminophenyl)metaborsäure Monohydrat, 3-Aminobenzolboronsäure Monohydrat

A18189.03 A18189.06
A18189.03EA 54 EUR
A18189.03 A18189.06
3-Aminophenylboronsäure Monohydrat 97%
3-Aminophenylboronsäure Monohydrat

Purity: 97% CAS No.: 206658-89-1 Molecular Formula: C6H8BNO2.H2O Formula Weight: 154.96 (136.95anhy) Melting Point: 93-97° Beilstein Reference: 2936342

Formel: C₆H₈BNO₂·1H₂O
Molecular Weight: 154.96 g/mol
Schmelzpunkt: 93…97 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149554
CAS-Nummer: 206658-89-1
EINECS: 250-189-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR