Samarium(III)bromid Hexahydrat 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Samariumtribromid Hexahydrat
A18191.09
A18191.09EA
53.9
EUR
A18191.09
Samarium(III)bromid Hexahydrat 99%
Samarium(III)bromid Hexahydrat
Purity: 99.99%
CAS No.: 13517-12-9
Molecular Formula: SmBr3.6H2O
Formula Weight: 498.15 (390.06anhy)
Melting Point:
Formel:
Br₃Sm·6H₂O Molecular Weight: 498,16 g/mol Schmelzpunkt: >640 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00192430 CAS-Nummer: 13517-12-9 EINECS: 237-347-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...