2-Amino-6-chlorpurin 99%
Lieferant: Thermo Fisher Scientific
Synonyme:
6-Chlor-7H-purin-2-ylamin
A18195.06
A18195.14
A18195.03
A18195.06EA
108
EUR
A18195.06
A18195.14
A18195.03
2-Amino-6-chlorpurin 99%
2-Amino-6-chlorpurin
Purity: 99%
CAS No.: 10310-21-1
Molecular Formula: C5H4ClN5
Formula Weight: 169.57
Melting Point:
Formel:
C₅H₄ClN₅ Molecular Weight: 169.57 g/mol Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075252 CAS-Nummer: 10310-21-1 EINECS: 233-686-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...