2-Amino-6-chlorpurin 99%

Lieferant: Thermo Fisher Scientific

Synonyme: 6-Chlor-7H-purin-2-ylamin

A18195.06 A18195.14 A18195.03
A18195.06EA 108 EUR
A18195.06 A18195.14 A18195.03
2-Amino-6-chlorpurin 99%
2-Amino-6-chlorpurin

Purity: 99% CAS No.: 10310-21-1 Molecular Formula: C5H4ClN5 Formula Weight: 169.57 Melting Point: 300° Beilstein Reference: 9626

Formel: C₅H₄ClN₅
Molecular Weight: 169.57 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00075252
CAS-Nummer: 10310-21-1
EINECS: 233-686-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR