2,4,6-Triphenylpyriliumtetrafluoroborat ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

A18217.06 A18217.14 A18217.22
A18217.06EA 59.4 EUR
A18217.06 A18217.14 A18217.22
2,4,6-Triphenylpyriliumtetrafluoroborat ≥97%
2,4,6-Triphenylpyriliumtetrafluoroborat

Purity: 97% CAS No.: 448-61-3 Molecular Formula: C23H17BF4O Formula Weight: 396.20 Melting Point: 248-254° Beilstein Reference: 3586533

Formel: C₂₃H₁₇BF₄O
Schmelzpunkt: 248…254 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012001
CAS-Nummer: 448-61-3
EINECS: 207-186-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR