2,4,6-Triphenylpyriliumtetrafluoroborat ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A18217.06
A18217.14
A18217.22
A18217.06EA
59.4
EUR
A18217.06
A18217.14
A18217.22
2,4,6-Triphenylpyriliumtetrafluoroborat ≥97%
2,4,6-Triphenylpyriliumtetrafluoroborat
Purity: 97%
CAS No.: 448-61-3
Molecular Formula: C23H17BF4O
Formula Weight: 396.20
Melting Point: 248-254°
Beilstein Reference: 3586533
Formel:
C₂₃H₁₇BF₄O Schmelzpunkt: 248…254 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012001 CAS-Nummer: 448-61-3 EINECS: 207-186-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...