n-Tridecan ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A18220.14
A18220.22
A18220.36
A18220.14EA
38.3
EUR
A18220.14
A18220.22
A18220.36
n-Tridecan ≥98%
n-Tridecan
Purity: 98+%
CAS No.: 629-50-5
Molecular Formula: C13H28
Formula Weight: 184.37
Flash Point: 79°(174°F)
Refractive Index: 1.4250
Melting Point: -5°
Boiling Point: 235°
Beilstein Reference: 1733089
MDL No.: MFCD00008979
Formel:
H₃C(CH₂)₁₁CH₃ Siedepunkt: 235 °C Schmelzpunkt: –5 °C Dichte: 0,755 Flammpunkt: 79 °C (174 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008979 CAS-Nummer: 629-50-5 EINECS: 211-093-4 UN: 3295 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...