Indium(III)nitrat Hydrat (≥29% In)

Lieferant: Thermo Scientific
Gefahr

Synonyme: Indium trinitrate hydrate

A18252.14
A18252.14EA 148 EUR
A18252.14
Indium(III)nitrat Hydrat (≥29% In)
Indium(III)nitrat Hydrat

CAS No.: 207398-97-8 Molecular Formula: In(NO3)3.xH2O Formula Weight: 300.83(anhy) Melting Point: 100° -H2O UN No.: 1477 MDL No.: MFCD00149742

Formel: InN₃O₉·H₂O
Molecular Weight: 318,85 g/mol
Schmelzpunkt: 100 °C (-H₂O)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149742
CAS-Nummer: 207398-97-8
EINECS: 237-393-5
UN: 1477
ADR: 5.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR