2,2,5,5-Tetramethyl-3-pyrroline-3-carboxamide 99%

Lieferant: Thermo Fisher Scientific
Achtung

A18294.03 A18294.06
A18294.03EA 31.7 EUR
A18294.03 A18294.06
2,2,5,5-Tetramethyl-3-pyrroline-3-carboxamide 99%
2,2,5,5-Tetramethyl-3-pyrroline-3-carboxamide

Purity: 99% CAS No.: 19805-75-5 Molecular Formula: C9H16N2O Formula Weight: 168.24 Melting Point: 179-183° MDL No.: MFCD00041846

Formel: C₉H₁₆N₂O
Schmelzpunkt: 179…183 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041846
CAS-Nummer: 19805-75-5
EINECS: 243-331-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR