Sudan III

Lieferant: Thermo Fisher Scientific

Synonyme: Sudan Rot, Sudan Red BK, Sudan® III (Rot), 1-(4-(Phenylazo)phenylazo)-2-naphthol, Tony Red, Scarlet B, 1-([4-[(Phenyl)diazenyl]phenyl]diazenyl)naphthalen-2-ol, Fat Ponceau G, Sudan G, Cerasin Red, Sudan Red B, Organol® Rot BS, Sudan Red III, Sudan V

A18318.22 A18318.14 A18318.36
A18318.22EA 56.2 EUR
A18318.22 A18318.14 A18318.36
Sudan III
Sudan III

CAS No.: 85-86-9 Molecular Formula: C22H16N4O Formula Weight: 352.40 Melting Point: ca 200° dec. Beilstein Reference: 931185 Merck Reference: 14,8884 MDL No.: MFCD00003905

Formel: C₂₂H₁₆N₄O
Molecular Weight: 352.4 g/mol
Schmelzpunkt: ∼ 200 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003905
CAS-Nummer: 85-86-9
EINECS: 201-638-4
Merck Index: 12,09054

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR