Ethyl-L-tyrosinathydrochlorid 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: H-Tyr-OEt·HCl, H-Tyr-OEt.HCl

A18358.14
A18358.14EA 55.9 EUR
A18358.14
Ethyl-L-tyrosinathydrochlorid 99%
Ethyl-L-tyrosinathydrochlorid

Purity: 99% CAS No.: 4089-07-0 Molecular Formula: C11H15NO3.HCl Formula Weight: 245.71 Melting Point: 166-168° Beilstein Reference: 4725904 Optical Rotation: -6.8° (c=2 in water) MDL No.: MFCD00063047

Formel: C₁₁H₁₅NO₃
Molecular Weight: 245,70 g/mol
Schmelzpunkt: 166…168 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00063047
CAS-Nummer: 4089-07-0
EINECS: 223-820-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR