Ethyl Rot
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A18367.09
A18367.18
A18367.30
A18367.09EA
34
EUR
A18367.09
A18367.18
A18367.30
Ethyl Rot
Ethyl red
CAS No.: 76058-33-8
Molecular Formula: C17H19N3O2
Formula Weight: 297.36
Melting Point: ca 135° dec.
Beilstein Reference: 3366056
MDL No.: MFCD00002427
Formel:
C₁₇H₁₉N₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002427 CAS-Nummer: 76058-33-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...