Cobalt(II)acetat Tetrahydrat ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

A18374.14 A18374.18 A18374.22 A18374.30 A18374.36
A18374.14EA 25.8 EUR
A18374.14 A18374.18 A18374.22 A18374.30 A18374.36
Cobalt(II)acetat Tetrahydrat ≥98%
Cobalt(II)acetat Tetrahydrat

Purity: 98% CAS No.: 6147-53-1 Molecular Formula: C4H6CoO4.4H2O Formula Weight: 249.08 (177.02anhy) Refractive Index: 1.542 Odour: Odourless Beilstein Reference: 3731397 Merck Reference: 14,2433 UN No.: 3077 MDL No.: MFCD00149569

Formel: (H₃CCOO)₂Co·4H₂O
Schmelzpunkt: ∼140 °C (-4H₂O)
Dichte: 1,71
Lagertemperatur: Raumtemperatur
MDL: MFCD00149569
CAS-Nummer: 6147-53-1
EINECS: 200-755-8
UN: 3077
ADR: 9,III
Merck Index: 14,2433

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR