D(+)-Tryptophan ≥99%

Lieferant: Thermo Fisher Scientific

A18426.06 A18426.14 A18426.22
A18426.06EA 37.7 EUR
A18426.06 A18426.14 A18426.22
D(+)-Tryptophan ≥99%
D(+)-Tryptophan

Purity: 99% CAS No.: 153-94-6 Molecular Formula: C11H12N2O2 Formula Weight: 204.23 Melting Point: 273° dec. Beilstein Reference: 86198

Formel: C₁₁H₁₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00064339
CAS-Nummer: 153-94-6
EINECS: 205-819-9
Merck Index: 14,09797

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR