Bromphenolblau

Lieferant: Thermo Fisher Scientific

Synonyme: 3,3',5,5'-Tetrabromphenolsulfonphthalein, 4,4'-(3H-2,1-Benzoxathiol-3-yliden)bis[2,6-dibromphenol]-S,S-dioxid

A18469.09 A18469.18
A18469.09EA 41.1 EUR
A18469.09 A18469.18
Bromphenolblau
Bromphenolblau

CAS No.: 115-39-9 Molecular Formula: C19H10Br4O5S Formula Weight: 669.98 Odour: Odourless Melting Point: ca 270° dec. Beilstein Reference: 61698 Merck Reference: 14,1444 MDL No.: MFCD00005875

Formel: C₁₉H₁₀Br₄O₅S
Schmelzpunkt: ∼ 270 °C (Zersetzung)
Dichte: 1 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005875
CAS-Nummer: 115-39-9
EINECS: 204-086-2
Merck Index: 14,1444

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR