2'-Hydroxy-6'-methoxyacetophenon ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-(2-Hydroxy-6-methoxyphenyl)ethanone

A18539.03 A18539.06 A18539.14
A18539.03EA 43.8 EUR
A18539.03 A18539.06 A18539.14
2'-Hydroxy-6'-methoxyacetophenon ≥97%
2'-Hydroxy-6'-methoxyacetophenon

Purity: 97% CAS No.: 703-23-1 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 56-59° Boiling Point: 141°/16mm MDL No.: MFCD00008732

Formel: C₉H₁₀O₃
Molecular Weight: 166.18 g/mol
Siedepunkt: 141 °C (16 mmHg)
Schmelzpunkt: 56…59 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008732
CAS-Nummer: 703-23-1
EINECS: 211-872-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR