2'-Hydroxy-6'-methoxyacetophenon ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1-(2-Hydroxy-6-methoxyphenyl)ethanone
A18539.03
A18539.06
A18539.14
A18539.03EA
43.8
EUR
A18539.03
A18539.06
A18539.14
2'-Hydroxy-6'-methoxyacetophenon ≥97%
2'-Hydroxy-6'-methoxyacetophenon
Purity: 97%
CAS No.: 703-23-1
Molecular Formula: C9H10O3
Formula Weight: 166.18
Melting Point: 56-59°
Boiling Point: 141°/16mm
MDL No.: MFCD00008732
Formel:
C₉H₁₀O₃ Molecular Weight: 166.18 g/mol Siedepunkt: 141 °C (16 mmHg) Schmelzpunkt: 56…59 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008732 CAS-Nummer: 703-23-1 EINECS: 211-872-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...