Salbutamol sulphate 99%

Lieferant: Thermo Fisher Scientific
Achtung

A18544.03 A18544.06
A18544.03EA 38.4 EUR
A18544.03 A18544.06
Salbutamol sulphate 99%
Salbutamol sulphate

Purity: 99% CAS No.: 51022-70-9 Molecular Formula: C26H42N2O6.H2SO4 Formula Weight: 576.70 MDL No.: MFCD00055200

Formel: (C₁₃H₂₁NO₃)₂·H₂SO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00055200
CAS-Nummer: 51022-70-9
EINECS: 256-916-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR