Ammoniumhydrogencarbonat 98
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Ammoniumbicarbonat
A18566.0E
A18566.36
A18566.0EEA
62
EUR
A18566.0E
A18566.36
Ammoniumhydrogencarbonat 98
Ammoniumhydrogencarbonat
Purity: 98%
CAS No.: 1066-33-7
Molecular Formula: (NH4)HCO3
Formula Weight: 79.06
Odour: Like ammonia
Melting Point: ca 60° dec.
Beilstein Reference: 4329606
Merck Reference: 14,497
MDL No.: MFCD00012138
Formel:
NH₄HCO₃ Molecular Weight: 79.06 g/mol Schmelzpunkt: ∼ 60 °C (Zers.) Dichte: 1.586 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012138 CAS-Nummer: 1066-33-7 EINECS: 213-911-5 Merck Index: 13,00497 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...