Nitrofurazon ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

A18593.14 A18593.22 A18593.30
A18593.14EA 24.6 EUR
A18593.14 A18593.22 A18593.30
Nitrofurazon ≥98%
Nitrofurazon

Purity: 98+% CAS No.: 59-87-0 Molecular Formula: C6H6N4O4 Formula Weight: 198.14 Melting Point: ca 240° dec. Beilstein Reference: 86403 MDL No.: MFCD00003225

Formel: C₆H₆N₄O₄
Schmelzpunkt: ∼ 240 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003225
CAS-Nummer: 59-87-0
EINECS: 200-443-1
Merck Index: 14,6600

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR