Uridine 5′-monophosphate disodium salt 99%
Lieferant: Thermo Fisher Scientific
A18601.06
A18601.14
A18601.22
A18601.06EA
84.5
EUR
A18601.06
A18601.14
A18601.22
Uridine 5′-monophosphate disodium salt 99%
Uridine 5′-monophosphate disodium salt
Purity: 99%
CAS No.: 3387-36-8
Molecular Formula: C9H11N2Na2O9P
Formula Weight: 368.15
Storage Temperature: 2°C to 8°C
Melting Point: ca 209° dec.
Beilstein Reference: 3582885
MDL No.: MFCD00006525
Formel:
C₉H₁₁N₂Na₂O₉P |
MDL:
MFCD00006525 CAS-Nummer: 3387-36-8 EINECS: 222-211-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...