8-Ethoxy-5-chinolinsulfonsäurehydrat 99%

Lieferant: Thermo Fisher Scientific
Achtung

A18622.18 A18622.09
A18622.18EA 176 EUR
A18622.18 A18622.09
8-Ethoxy-5-chinolinsulfonsäurehydrat 99%
8-Ethoxy-5-chinolinsulfonsäurehydrat

Purity: 99% CAS No.: 15301-40-3 Molecular Formula: C11H11NO4S.H2O Formula Weight: 271.29 (253.28anhy) Melting Point: ca 296° dec. Beilstein Reference: 225878 MDL No.: MFCD00065059

Formel: C₁₁H₁₁NO₄S
Schmelzpunkt: ∼ 296 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00065059
CAS-Nummer: 15301-40-3
EINECS: 239-334-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR