8-Ethoxy-5-chinolinsulfonsäurehydrat 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A18622.18
A18622.09
A18622.18EA
176
EUR
A18622.18
A18622.09
8-Ethoxy-5-chinolinsulfonsäurehydrat 99%
8-Ethoxy-5-chinolinsulfonsäurehydrat
Purity: 99%
CAS No.: 15301-40-3
Molecular Formula: C11H11NO4S.H2O
Formula Weight: 271.29 (253.28anhy)
Melting Point: ca 296° dec.
Beilstein Reference: 225878
MDL No.: MFCD00065059
Formel:
C₁₁H₁₁NO₄S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00065059 CAS-Nummer: 15301-40-3 EINECS: 239-334-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...