1-Brom-2,5-difluor-4-nitrobenzol ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
4-Brom-2,5-difluornitrobenzol
A18670.06
A18670.06EA
192
EUR
A18670.06
1-Brom-2,5-difluor-4-nitrobenzol ≥97%
1-Bromo-2,5-difluoro-4-nitrobenzene
Purity: 97%
CAS No.: 167415-27-2
Molecular Formula: C6H2BrF2NO2
Formula Weight: 237.99
Melting Point: 55-57°
Beilstein Reference: 7411347
Formel:
C₆H₂BrF₂NO₂ Schmelzpunkt: 55…57 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042184 CAS-Nummer: 167415-27-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...