2,3-Dihydro-1,4-benzodioxin-6-carbaldehyd 99%

Lieferant: Thermo Scientific

Synonyme: 2,3-Dihydro-1,4-benzodioxine-6-carbaldehyde, 1,4-Benzodioxan-6-carboxaldehyde, 2,3-dihydrobenzo[b][1,4]dioxine-6-carbaldehyde

A18696.06
A18696.06EA 51.1 EUR
A18696.06
2,3-Dihydro-1,4-benzodioxin-6-carbaldehyd 99%
2,3-Dihydrobenzo[b][1,4]dioxine-6-carbaldehyde

Purity: 99% CAS No.: 29668-44-8 Molecular Formula: C9H8O3 Formula Weight: 164.16 Flash Point: 110°(230°F) Melting Point: 48-52° Boiling Point: 104-106°/15mm MDL No.: MFCD00010092

Formel: C₉H₈O₃
Molecular Weight: 164,16 g/mol
Siedepunkt: 104…106 °C (15 mmHg)
Schmelzpunkt: 48…52 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010092
CAS-Nummer: 29668-44-8
EINECS: 249-763-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR