Dipropylenglycol (Isomerengemisch) ≥99%
Lieferant: Thermo Fisher Scientific
Purity: 99%
CAS No.: 25265-71-8
Molecular Formula: HOC3H6OC3H6OH
Formula Weight: 134.18
Physical Form: Liquid
Density: 1.022
Refractive Index: 1.4410
Sensitivity: Hygroscopic
Appearance: Colourless
Odour: Odourless
Melting Point: -32°
Boiling Point: 231-235°
MDL No.: MFCD00051023
Formel:
C₆H₁₄O₃ Siedepunkt: 231…235 °C Schmelzpunkt: –32 °C Dichte: 1,022 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051023 CAS-Nummer: 25265-71-8 EINECS: 246-770-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...