1-(2-Naphthoyl)-3,3,3-trifluoraceton ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4,4,4-Trifluoro-1-(naphthalen-2-yl)butane-1,3-dione, 4,4,4-Trifluoro-1-(2-naphthyl)-1,3-butanedione
A18726.06
A18726.14
A18726.06EA
33.4
EUR
A18726.06
A18726.14
1-(2-Naphthoyl)-3,3,3-trifluoraceton ≥99%
1-(2-Naphthoyl)-3,3,3-trifluoraceton
Purity: 99%
CAS No.: 893-33-4
Molecular Formula: C14H9F3O2
Formula Weight: 266.22
Melting Point: 74-76°
MDL No.: MFCD00054501
Formel:
C₁₄H₉F₃O₂ Molecular Weight: 266.22 g/mol Schmelzpunkt: 74…76 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00054501 CAS-Nummer: 893-33-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...