Nerolidol (cis-/trans-Gemisch) ≥97%

Lieferant: Thermo Fisher Scientific

A18812.14 A18812.22
A18812.14EA 32.7 EUR
A18812.14 A18812.22
Nerolidol (cis-/trans-Gemisch) ≥97%
Nerolidol (cis-/trans-Gemisch)

Purity: 97+% CAS No.: 7212-44-4 Molecular Formula: C15H26O Formula Weight: 222.37 Flash Point: 96°(204°F) Refractive Index: 1.4790 Boiling Point: 263-266° Beilstein Reference: 1724135 MDL No.: MFCD00008911

Formel: C₁₅H₂₆O
Siedepunkt: 263…266 °C
Dichte: 0,876
Flammpunkt: 96 °C (204 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008911
CAS-Nummer: 7212-44-4
EINECS: 230-597-5
Merck Index: 14,6477

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR