D(-)-Valin 98
Lieferant: Thermo Fisher Scientific
Synonyme:
(R)-(-)-Valin, (R)-(-)-2-Amino-3-methylbutansäure, (R)-(-)-2-Aminoisovaleriansäure, H-D-Val-OH
A18894.06
A18894.14
A18894.22
A18894.06EA
36.1
EUR
A18894.06
A18894.14
A18894.22
D(-)-Valin 98
D(-)-Valin
Purity: 98+%
CAS No.: 640-68-6
Molecular Formula: C5H11NO2
Formula Weight: 117.15
Melting Point: 295° dec.
Beilstein Reference: 1721135
Formel:
C₅H₁₁NO₂ Molecular Weight: 117.15 g/mol Siedepunkt: 217 Schmelzpunkt: 295 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064219 CAS-Nummer: 640-68-6 EINECS: 211-368-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...