Boc-ON ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A18906.06
A18906.14
A18906.22
A18906.06EA
31.6
EUR
A18906.06
A18906.14
A18906.22
Boc-ON ≥98%
Boc-ON
Purity: 98+%
CAS No.: 58632-95-4
Molecular Formula: C13H14N2O3
Formula Weight: 246.27
Storage Temperature: 2°C to 8°C
Melting Point: 86-89°
Beilstein Reference: 2217296
UN No.: UN3439
MDL No.: MFCD00001863
Formel:
C₁₃H₁₄N₂O₃ Schmelzpunkt: 86…89 °C |
MDL:
MFCD00001863 CAS-Nummer: 58632-95-4 EINECS: 261-370-9 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...