Thionin (Acetat)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3,7-Diaminophenothiazin-5-ium acetate
A18912.06
A18912.14
A18912.06EA
56.2
EUR
A18912.06
A18912.14
Thionin (Acetat)
Thionin (Acetat)
CAS No.: 78338-22-4
Molecular Formula: C14H13N3O2S
Formula Weight: 287.34
MDL No.: MFCD00081194
Used as a biogical stain
Formel:
C₁₄H₁₃N₃O₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00081194 CAS-Nummer: 78338-22-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...