Thionin (Acetat)

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3,7-Diaminophenothiazin-5-ium acetate

A18912.06 A18912.14
A18912.06EA 56.2 EUR
A18912.06 A18912.14
Thionin (Acetat)
Thionin (Acetat)

CAS No.: 78338-22-4 Molecular Formula: C14H13N3O2S Formula Weight: 287.34 MDL No.: MFCD00081194


Used as a biogical stain

Formel: C₁₄H₁₃N₃O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00081194
CAS-Nummer: 78338-22-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR