3-Mercapto-1,2-propandiol ≥90%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A18948.18
A18948.30
A18948.18EA
59.3
EUR
A18948.18
A18948.30
3-Mercapto-1,2-propandiol ≥90%
3-Mercapto-1,2-propandiol
Purity: 90+%
CAS No.: 96-27-5
Molecular Formula: C3H8O2S
Formula Weight: 108.16
Flash Point: 149°(300°F)
Refractive Index: 1.5110
Boiling Point: 117-119°/5mm
Beilstein Reference: 1732046
Merck Reference: 14,9335
MDL No.: MFCD00004879
Formel:
C₃H₈O₂S Siedepunkt: 117…119 °C (5 mmHg) Dichte: 1,247 Flammpunkt: 149 °C (300 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004879 CAS-Nummer: 96-27-5 EINECS: 202-495-0 Merck Index: 14,9335 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...