3'-Fluor-4'-methoxyacetophenon ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

A18972.06 A18972.14
A18972.06EA 28.4 EUR
A18972.06 A18972.14
3'-Fluor-4'-methoxyacetophenon ≥99%
3'-Fluor-4'-methoxyacetophenon

Purity: 99% CAS No.: 455-91-4 Molecular Formula: C9H9FO2 Formula Weight: 168.17 Melting Point: 90-94° Boiling Point: 147-148°/20mm MDL No.: MFCD00026219

Formel: C₉H₉FO₂
Siedepunkt: 147…148 °C (20 mmHg)
Schmelzpunkt: 90…94 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00026219
CAS-Nummer: 455-91-4
EINECS: 207-253-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR