Phosphonitrilic chloride trimer 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A18986.22
A18986.06
A18986.14
A18986.22EA
281
EUR
A18986.22
A18986.06
A18986.14
Phosphonitrilic chloride trimer 98%
Phosphonitrilic chloride trimer
Purity: 98%
CAS No.: 940-71-6
Molecular Formula: Cl6N3P3
Formula Weight: 347.66
Melting Point: 111-115°
Boiling Point: 127°/13mm
UN No.: UN3260
MDL No.: MFCD00006474
Formel:
Cl₆N₃P₃ Siedepunkt: 127 °C (13 mmHg) Schmelzpunkt: 111…115 °C Dichte: 1,98 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006474 CAS-Nummer: 940-71-6 EINECS: 213-376-8 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...