5-Ioduracil ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

A18994.09 A18994.18
A18994.09EA 51.5 EUR
A18994.09 A18994.18
5-Ioduracil ≥97%
5-Ioduracil

Purity: 97% CAS No.: 696-07-1 Molecular Formula: C4H3IN2O2 Formula Weight: 237.99 Melting Point: 274-276° Beilstein Reference: 4891

Formel: C₄H₃IN₂O₂
Siedepunkt: 401 °C (1013 hPa)
Schmelzpunkt: 274…276 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006020
CAS-Nummer: 696-07-1
EINECS: 211-788-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR