cis-3-Octen-1-ol 95%
Lieferant: Thermo Fisher Scientific
Achtung
|
A19014.09
A19014.18
A19014.09EA
41.4
EUR
A19014.09
A19014.18
cis-3-Octen-1-ol 95%
cis-3-Octen-1-ol
Purity: 95%
CAS No.: 20125-84-2
Molecular Formula: C8H16O
Formula Weight: 128.22
Flash Point: 82°(179°F)
Refractive Index: 1.4470
Boiling Point: 88-89°/15mm
Beilstein Reference: 1720075
MDL No.: MFCD00014062
Formel:
C₈H₁₆O Siedepunkt: 88…89 °C (15 mmHg) Dichte: 0,850 Flammpunkt: 82 °C (179 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014062 CAS-Nummer: 20125-84-2 EINECS: 243-533-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...