Butylmethacrylat 99% (durch GC) stabilisiert

Lieferant: Thermo Scientific
Achtung

Synonyme: Methacrylsäurebutylester, 2-Methylpropensäurebutylester

Stabilisierung: Stabilisiert mit Hydrochinonmonomethylether (4-Methoxyphenol, MEHQ)

A19068.AP
A19068.APEA 32.8 EUR
A19068.AP
Butylmethacrylat 99% (durch GC) stabilisiert
Butylmethacrylat

Stabilized with 15-20 ppm 4-methoxyphenol


n-Butyl methacrylate is used as a building block of polymer based products that are used in paints and coatings, resins, adhesives, toners and inks, oil additives for dental and medical products. Its polymer polybutyl methacrylate (PBMA) with the sodium salt of partially carboxymethylated guar gum (Na-PCMGG, DS = 0.497) is synthesized using ceric ammonium nitrate as a redox initiator.


Bitte beachten Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents. Store in a cool place.

Formel: C₈H₁₄O₂
Molecular Weight: 142,2 g/mol
Siedepunkt: 160…163 °C
Schmelzpunkt: –75 °C
Dichte: 0,894
Flammpunkt: 49 °C (120 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00009444
CAS-Nummer: 97-88-1
EINECS: 202-615-1
UN: 2227
ADR: 3,III

Bestellen

Spezifikation Testergebnisse

Appearance (colour) Clear, colourless to pale yellow
Form Liquid
Assay (GC) >98.5%
Refractive Index 1.4225 - 1.4255
Solution Test 10% solution in methanol - clear

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR