Methyl-4-brom-3-methylbenzoat ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Methyl 4-bromo-3-methylbenzoate

A19078.06 A19078.14
A19078.06EA 60.9 EUR
A19078.06 A19078.14
Methyl-4-brom-3-methylbenzoat ≥97%
Methyl-4-brom-3-methylbenzoat

Purity: 97% CAS No.: 148547-19-7 Molecular Formula: C9H9BrO2 Formula Weight: 229.07 Melting Point: 39-44° Beilstein Reference: 6593534

Formel: C₉H₉BrO₂
Molecular Weight: 229.07 g/mol
Schmelzpunkt: 39…44 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00673014
CAS-Nummer: 148547-19-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR