3-Nonen-2-on 96%
Lieferant: Thermo Fisher Scientific
Achtung
|
Synonyme:
Non-3-en-2-one
A19084.09
A19084.18
A19084.30
A19084.09EA
34.5
EUR
A19084.09
A19084.18
A19084.30
3-Nonen-2-on 96%
3-Nonen-2-on
Purity: 96%
CAS No.: 14309-57-0
Molecular Formula: C9H16O
Formula Weight: 140.23
Flash Point: 81°(177°F)
Refractive Index: 1.4490
Boiling Point: 85°/12mm
Beilstein Reference: 1743558
MDL No.: MFCD00010241
Formel:
C₉H₁₆O Siedepunkt: 85 °C (12 mmHg) Dichte: 0,848 Flammpunkt: 81 °C (177 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010241 CAS-Nummer: 14309-57-0 EINECS: 238-248-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...