O-Methylhydroxylamin Hydrochlorid ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A19188.06
A19188.14
A19188.06EA
87.4
EUR
A19188.06
A19188.14
O-Methylhydroxylamin Hydrochlorid ≥98%
O-Methylhydroxylamin Hydrochlorid
Purity: 98+%
CAS No.: 593-56-6
Molecular Formula: CH3ONH2.HCl
Formula Weight: 83.52
Melting Point: ca 148° dec.
Beilstein Reference: 3589723
Merck Reference: 14,5989
UN No.: 3261
MDL No.: MFCD00012951
Formel:
CH₆ClNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012951 CAS-Nummer: 593-56-6 EINECS: 209-798-7 UN: 3261 ADR: 8,III Merck Index: 14,5989 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...