3-Methyl-2-thenoesäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A19194.06 A19194.14 A19194.22
A19194.06EA 39.8 EUR
A19194.06 A19194.14 A19194.22
3-Methyl-2-thenoesäure ≥98%
3-Methyl-2-thenoesäure

Purity: 98% CAS No.: 23806-24-8 Molecular Formula: C6H6O2S Formula Weight: 142.18 Melting Point: 144-149° Beilstein Reference: 116184

Formel: C₆H₆O₂S
Schmelzpunkt: 144…149 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005438
CAS-Nummer: 23806-24-8
EINECS: 245-894-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR