2'-Hydroxy-5'-methoxyacetophenon ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2'-Hydroxy-5'-Methoxyacetophenone, 2′-Hydroxy-5′-methoxyacetophenone
A19216.06
A19216.14
A19216.06EA
58.6
EUR
A19216.06
A19216.14
2'-Hydroxy-5'-methoxyacetophenon ≥99%
2'-Hydroxy-5'-methoxyacetophenon
Purity: 99%
CAS No.: 705-15-7
Molecular Formula: C9H10O3
Formula Weight: 166.18
Flash Point:
Formel:
C₉H₁₀O₃ Molecular Weight: 166.18 g/mol Siedepunkt: 281 Schmelzpunkt: 48…50 °C Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008731 CAS-Nummer: 705-15-7 EINECS: 211-882-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...