5-Brom-2-thenoesäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
5-Bromthiophen-2-carbonsäure, 5-Brom-2-thiophencarbonsäure
A19242.03
A19242.06
A19242.14
A19242.03EA
28.6
EUR
A19242.03
A19242.06
A19242.14
5-Brom-2-thenoesäure ≥98%
5-Brom-2-thenoesäure
Purity: 98%
CAS No.: 7311-63-9
Molecular Formula: C5H3BrO2S
Formula Weight: 207.05
Melting Point: 139-143°
Beilstein Reference: 118364
Formel:
C₅H₃BrO₂S Schmelzpunkt: 139…143 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00079725 CAS-Nummer: 7311-63-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...